Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Furan-2-acetic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA458390010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS459-72-3
IUPAC Nameethyl 2-fluoroacetate
Molecular FormulaC4H7FO2
InChI KeyVCYZVXRKYPKDQB-UHFFFAOYSA-N
SMILESCCOC(=O)CF
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%