Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
CAS: 120011-70-3 | C24H30ClNO3 | 415.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA458050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDonepezil hydrochloride
CAS120011-70-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/