Indomethacin
Indomethacin
Indomethacin
Thermo Scientific Chemicals
Indomethacin
Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID).
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number FSA458030250
Price (MYR)
491.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
491.00
Add to cart
Specifications
Chemical Name or MaterialIndomethacin
CAS53-86-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
May cause an allergic skin reaction.
May damage fertility.
May damage the unborn child.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use
View more
General Description
• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release
• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics
• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation
Applications
• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions
• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin
RUO – Research Use Only
General References:
  1. Mitra, S.; Florez, I.D.; Tamayo, M.E.; Mbuagbaw, L.; Vanniyasingam, T.; Veroniki, A.A.; Zea, A.M.; Zhang, Y.; Sadeghirad, B.; Thabane, L. Association of Placebo, Indomethacin, Ibuprofen, and Acetaminophen With Closure of Hemodynamically Significant Patent Ductus Arteriosus in Preterm Infants: A Systematic Review and Meta-analysis. JAMA. 2018, 319(12), 1221-1238.
  2. El-Mashad, A.E.; El-Mahdy, H.; El Amrousy, D.; Elgendy, M. Comparative study of the efficacy and safety of paracetamol, ibuprofen, and indomethacin in closure of patent ductus arteriosus in preterm neonates. Eur J Pediatr. 2017, 176(2), 233-240.