Finasteride, 98%
Finasteride, 98%
Finasteride, 98%
Thermo Scientific Chemicals
Finasteride, 98%
Finasteride, CAS # 98319-26-7, is a 5α-reductase inhibitor that blocks conversion of testosterone and inhibits neurosteroid production.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA458000010
Price (MYR)
1,267.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,267.00
Add to cart
Specifications
Chemical Name or MaterialFinasteride
CAS98319-26-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
May damage the unborn child.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Obtain special instructions before use.
Use personal protective equipment as
View more
General Description
  • Finasteride is an inhibitor of the enzyme 5α-reductase, which converts testosterone to dihydrotestosterone (DHT)
  • Finasteride also has been explored in neuroscience, as 5α-reductase enzymes are present in the central nervous system
  • Finasteride has been seen to cross the blood-brain barrier and block production of neurosteroids
Applications
  • Finasteride is mainly used in studies of prostate hyperplasia and tumors in mice, and can be studied in vitro or in vivo for its effects on hyperplasia, proliferation, and antioxidant activity
  • Its activity has also been explored in brain, liver and testis tissues
  • In vivo injection of finasteride has been shown to decrease neurogenesis and influence neuronal plasticity in mice
RUO – Research Use Only
General References:
  1. Kim, H.J.; Kim, T.J.; Kim, Y.G.; Seong, C.; Cho, J.H.; Kim, W.; Lee, K.H.; Kim, D.Y. Antioxidant and Antiproliferative Activity of Finasteride against Glioblastoma Cells. Pharmaceutics. 2021, 13(9), 1410.
  2. Chhipa, R.R.; Halim, D.; Cheng, J.; Zhang, H.Y.; Mohler, J.L.; Ip, C.; Wu, Y. The direct inhibitory effect of dutasteride or finasteride on androgen receptor activity is cell line specific. Prostate. 2013, 73(14), 1483-94.
  3. Römer, B.; Pfeiffer, N.; Lewicka, S.; Ben-Abdallah, N.; Vogt, M.A.; Deuschle, M.; Vollmayr, B.; Gass, P. Finasteride treatment inhibits adult hippocampal neurogenesis in male mice. Pharmacopsychiatry. 20100, 43(5), 174-8.