2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™
2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™
2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™
2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™
2,7-Dibromo-9,9-dimethylfluorene, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA457310050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2,7-Dibromo-9,9-dimethylfluorene
CAS28320-32-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.