Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
CAS: 95058-81-4 | C9H11F2N3O4 | 263.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA456890010
Price (MYR)
612.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS50-89-5
IUPAC Name1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC10H14N2O5
InChI KeyIQFYYKKMVGJFEH-UHFFFAOYNA-N
SMILESCC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
UV spectrophotometry98.0 to 102.0 % (lambda max. 267 nm, pH 7)
Water=<0.5 % (K.F.)
Infrared spectrumConforms
HPLC>=99.0 %