Salbutamol hemisulfate, 98%
Salbutamol hemisulfate, 98%
Salbutamol hemisulfate, 98%
Salbutamol hemisulfate, 98%
Thermo Scientific Chemicals
Salbutamol hemisulfate, 98%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA456460010
Price (MYR)
638.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
638.00
Add to cart
Specifications
Chemical Name or MaterialSalbutamol hemisulfate
CAS51022-70-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Suspected of causing cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.