Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number FSA456290250
Price (MYR)
2,749.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,749.00
Add to cart
Chemical Identifiers
CAS112780-04-8
IUPAC Namemethyl 4-hydroxyhex-2-ynoate
Molecular FormulaC7H10O3
InChI KeyGMESNUCOCRRYJV-UHFFFAOYNA-N
SMILESCCC(O)C#CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.4600-1.4650 @ 20?C