Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number FSA456290050
Price (MYR)
741.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
741.00
Add to cart
Chemical Identifiers
CAS121-69-7
IUPAC NameN,N-dimethylaniline
Molecular FormulaC8H11N
InChI KeyJLTDJTHDQAWBAV-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5570 to 1.5590 (20°C, 589 nm)