Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number FSA456290010
Price (MYR)
404.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
404.00
Add to cart
Chemical Identifiers
CAS555-44-2
IUPAC Name1,3-bis(hexadecanoyloxy)propan-2-yl hexadecanoate
Molecular FormulaC51H98O6
InChI KeyPVNIQBQSYATKKL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to almost white
Infrared spectrumConforms
HPLC>=98.5 %