Docetaxel, 98%
Docetaxel, 98%
Docetaxel, 98%
Thermo Scientific Chemicals
Docetaxel, 98%
CAS: 114977-28-5 | C43H53NO14 | 807.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number FSA456262500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS10373-78-1
IUPAC Name1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Molecular FormulaC10H14O2
InChI KeyVNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(=O)C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)199-206?C
Assay (GC)≥98.5%