Oxaliplatin, 99%
Oxaliplatin, 99%
Oxaliplatin, 99%
Thermo Scientific Chemicals
Oxaliplatin, 99%
CAS: 61825-94-3 | C8H14N2O4Pt | 397.29 g/mol
Have Questions?
Catalog NumberQuantity
FSA456131000100 mg
Catalog number FSA456131000
Price (MYR)
1,672.00
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
1,672.00
Add to cart
Chemical Identifiers
CAS830-03-5
IUPAC Name4-nitrophenyl acetate
Molecular FormulaC8H7NO4
InChI KeyQAUUDNIGJSLPSX-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point76°C to 79°C
GC>=96.0 %