Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Catalog number FSA456071000
Price (MYR)
62.53
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
62.53
Add to cart
Chemical Identifiers
CAS121-90-4
IUPAC Name3-nitrobenzoyl chloride
Molecular FormulaC7H4ClNO3
InChI KeyNXTNASSYJUXJDV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Melting point30°C to 35°C
Infrared spectrumConforms
Titration RCOCl>=97.5 %
Appearance (Form)Low melting solid