Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Catalog number FSA456070050
Price (MYR)
636.74
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
636.74
Add to cart
Specifications
Chemical Name or MaterialMontmorillonite K-10
CAS1318-93-0
Quantity5 kg
EINECS Number215-288-5
Molecular FormulaAl2H2O12Si4