Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
5 kg
Catalog number FSA456070010
Price (MYR)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialMontmorillonite K-10
CAS1318-93-0
Quantity1 kg
EINECS Number215-288-5
Molecular FormulaAl2H2O12Si4