Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA455830010
Price (MYR)
509.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
509.00
Add to cart
Chemical Identifiers
CAS10424-65-4
IUPAC Nametetramethylazanium pentahydrate hydroxide
Molecular FormulaC4H23NO6
InChI KeyMYXKPFMQWULLOH-UHFFFAOYSA-M
SMILESO.O.O.O.O.[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point68°C to 73°C
Carbonate (CO3)=<7500 ppm
Impurity=<0.1 % Halides (as Cl)