Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
CAS: 497-76-7 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA455640050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1608-26-0
IUPAC Name[bis(dimethylamino)phosphanyl]dimethylamine
Molecular FormulaC6H18N3P
InChI KeyXVDBWWRIXBMVJV-UHFFFAOYSA-N
SMILESCN(C)P(N(C)C)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4630-1.4680 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%