Fluconazole, 98%
Fluconazole, 98%
Fluconazole, 98%
Fluconazole, 98%
Fluconazole, 98%, CAS # 86386-73-4, is a synthetic triazole with antifungal activity against some species of Candida. It also shows inhibitory activity against cytochrome P-450 enzymes and it is a demethylase.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA455480010
Price (MYR)
660.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
660.00
Add to cart
Chemical Identifiers
CAS101-61-1
IUPAC Name4-{[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Molecular FormulaC17H22N2
InChI KeyJNRLEMMIVRBKJE-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(CC2=CC=C(C=C2)N(C)C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or brown or blue to purple to gray
Assay (GC)≥98.0%
FormCrystals or powder or crystalline powder or fused solid or flakes
Melting Point (clear melt)84.0-90.0°C
Identification (FTIR)Conforms
General Description
• Fluconazole, 98% is a triazole compound showing antifungal activity mainly against Candida, Coccidioides, and Cryptococcus species
• It has been shown to inhibit fungal cytochrome P-450 sterol C-14 α-demethylation, resulting in the accumulation of fungal 14 α-methyl sterols, the loss of normal fungal sterols, and associated fungistatic activity
• It also inhibits cytochrome P450 family 2 subfamily c member 9 (CYP2C9)
• Mammalian cell demethylation is much less susceptible to fluconazole inhibition
Applications
• Fluconazole has fungistatic activity against many fungal strains including Candida albicans, Candida glabrata, Candida parapsilosis, Candida tropicalis, and Cryptococcus neoformans.
• Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene
• In biological experiments, this compound has been used to assess minimum inhibitory concentration in (MIC)-fungal assays
• It has also been used as a positive control in antifungal assay of plants
RUO – Research Use Only
General References:
  1. Clancy, C.J.; Yu, V.L.; Morris, A.J.; Snydman, D.R.; Nguyen, M.H. Fluconazole MIC and the fluconazole dose/MIC ratio correlate with therapeutic response among patients with candidemia. Antimicrob Agents Chemother. 2005, 49 (8), 3171-3177.
  2. Matsui, K.; Tsume, Y.; Amidon, G. E.; Amidon, G. L. In Vitro Dissolution of Fluconazole and Dipyridamole in Gastrointestinal Simulator (GIS), Predicting in Vivo Dissolution and Drug-Drug Interaction Caused by Acid-Reducing Agents. Mol. Pharmaceutics. 2015, 12 (7), 2418-2428.
  3. Bhardwaj, T.; Bhardwaj, V.; Sharma, K.; Gupta, A.; Cameotra, S. S.; Sharma, P. Thermo-acoustical analysis of sodium dodecyl sulfate: Fluconazole (antifungal drug) based micellar system in hydro-ethanol solutions for potential drug topical application. J. Chem. Thermodyn. 2014, 78, 1-6.