Clotrimazole
Clotrimazole
Clotrimazole
Clotrimazole
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA455461000
Price (MYR)
5,184.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
5,184.00
Add to cart
Chemical Identifiers
CAS552-86-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown or yellow-orange
FormPowder, crystals, crystalline powder and/or chunks
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Melting Point (clear melt)133-142?C