Clotrimazole
Clotrimazole
Clotrimazole
Clotrimazole
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number FSA455460050
Price (MYR)
413.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
413.00
Add to cart
Specifications
Chemical Name or MaterialClotrimazole
CAS23593-75-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con