Oxacillin sodium salt, 95%
Oxacillin sodium salt, 95%
Oxacillin sodium salt, 95%
Oxacillin sodium salt, 95%
Oxacillin sodium salt, 95%
Oxacillin sodium salt, CAS # 1173-88-2, is an antibiotic that is used primarily in studying gram-positive bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA455440010
Price (MYR)
350.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
350.00
Add to cart
Specifications
Chemical Name or MaterialOxacillin sodium salt
TypeOxacillin Sodium Salt
CAS1173-88-2
Infrared SpectrumAuthentic
Water3.5 to 5.0%
View more
General Description
  • Oxacillin sodium salt is a narrow-spectrum beta-lactam antibiotic
  • It inhibits biosynthesis of bacterial cell walls at the level of transpeptidation (PBP) of peptidoglycans
Applications
  • Oxacillin sodium salt is an antibiotic that is active against gram-positive bacteria
  • Oxacillin can be used in studies of mechanisms of resistance to penicillinase and antimicrobial susceptibility
  • It has also been used to study autolysins
RUO – Research Use Only
General References:
  1. Qiao, Y.; Srisuknimit, V.; Rubino, F.; et al. Lipid II overproduction allows direct assay of transpeptidase inhibition by β-lactams. Nat Chem Biol. 2017, 13(7), 793-798.
  2. Best, G.K.; Best, N.H.; Koval, A.V. Evidence for participation of autolysins in bactericidal action of oxacillin on Staphylococcus aureus. Antimicrob Agents Chemother. 1974, 6(6), 825-30.