Clarithromycin, 97%
Clarithromycin, 97%
Clarithromycin, 97%
Clarithromycin, 97%
CAS: 81103-11-9 | C38H69NO13 | 747.95 g/mol
Have Questions?
Catalog NumberQuantity
FSA4552200101 g
Catalog number FSA455220010
Price (MYR)
1,374.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,374.00
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point132°C to 135°C
Appearance (Form)Crystals
Infrared spectrumConforms
Titration with HClO499.0 to 100.5 %
Heavy metals (as Pb)=<10 ppm