4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl phosphate, 97%, Thermo Scientific Chemicals
CAS: 330-13-2 | C6H6NO6P | 219.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA455010100
Price (MYR)
725.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
725.00
Add to cart
Specifications
Chemical Name or Material4-Nitrophenyl phosphate
CAS330-13-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.