4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, 97%
CAS: 81-81-2 | C19H16O4 | 308.34 g/mol
Have Questions?
Catalog NumberQuantity
FSA45468010010 g
Catalog number FSA454680100
Price (MYR)
561.58
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
561.58
Add to cart
Chemical Identifiers
CAS36854-57-6
IUPAC Name2-phenylbutanoyl chloride
Molecular FormulaC10H11ClO
InChI KeyQGXMHCMPIAYMGT-UHFFFAOYNA-N
SMILESCCC(C(Cl)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5150-1.5190 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%