Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
Alfa Aesar
Decamethylcyclopentasiloxane, 97%, Thermo Scientific Chemicals
CAS: 541-02-6 | C10H30O5Si5 | 370.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number FSA454580500
Price (MYR)
323.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
323.00
Add to cart
Specifications
Chemical Name or MaterialDecamethylcyclopentasiloxane
CAS541-02-6
Density0.9590g/mL
EINECS Number208-764-9
Boiling Point101.0°C (20.0 mmHg)