Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
Barium nitrate, pure, Thermo Scientific Chemicals
CAS: 10022-31-8 | BaN2O6 | 261.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA453805000
Price (MYR)
287.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
287.00
Add to cart
Chemical Identifiers
CAS590-29-4
IUPAC Namepotassium formate
Molecular FormulaCHKO2
InChI KeyWFIZEGIEIOHZCP-UHFFFAOYSA-M
SMILES[K+].[O-]C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Loss on Drying<2.0%
Identification (FTIR)Conforms
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)161-172?C