Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Periodic acid, Thermo Scientific Chemicals
Alfa Aesar
Periodic acid, Thermo Scientific Chemicals
CAS: 10450-60-9 | H5IO6 | 227.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA453170250
Price (MYR)
398.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
398.00
Add to cart
Specifications
Chemical Name or MaterialPeriodic Acid
CAS10450-60-9
Melting Point122.0°C
Quantity25 g
Molecular FormulaH5IO6