Perchloric acid, Thermo Scientific Chemicals
Perchloric acid, Thermo Scientific Chemicals
Perchloric acid, Thermo Scientific Chemicals
Perchloric acid, Thermo Scientific Chemicals
Alfa Aesar
Perchloric acid, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number FSA452850010
Price (MYR)
2,554.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
2,554.00
Add to cart
Chemical Identifiers
CAS105-45-3
IUPAC Namemethyl 3-oxobutanoate
Molecular FormulaC5H8O3
InChI KeyWRQNANDWMGAFTP-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (Silylated GC)≥98.5%
Refractive Index1.4175-1.4215 @ 20?C