1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals
CAS: 84322-56-5 | C6F3I3 | 509.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA452690010
Price (MYR)
840.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
840.00
Add to cart
Chemical Identifiers
CAS122-97-4
IUPAC Name3-phenylpropan-1-ol
Molecular FormulaC9H12O
InChI KeyVAJVDSVGBWFCLW-UHFFFAOYSA-N
SMILESOCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.5255-1.5285 @ 20?C