Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetamide, 99%, for analysis, Thermo Scientific Chemicals
CAS: 60-35-5 | C2H5NO | 59.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number FSA451872500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES[H+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Strength0.998 to 1.002 N (20°C)