3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
3-Methyl-1-butanol, for analysis
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantity
FSA4516600101 L
Catalog number FSA451660010
Price (MYR)
190.90
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
190.90
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gold
FormRod, 3mm (0.1in) diameter.
Assay from Supplier's CofA≥99.9985% (metals basis)
Length≈3.7cm