3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Oxetanone, 95%, Thermo Scientific Chemicals
CAS: 6704-31-0 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA451200050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS8006-28-8
IUPAC Namesodium oxocalcium hydroxide
Molecular FormulaCaHNaO2
InChI KeyHUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES[OH-].[Na+].O=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
CO2 absorption capacity>=19.0 %
IndicatorPasses test
Appearance (Color)White to beige to light grey
Loss on drying=<20 % (10 g, 200°C, 2 h)