Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Sulfuric acid, for analysis, 25% solution in water
Thermo Scientific Chemicals
Sulfuric acid, for analysis, 25% solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
FSA4510500101 L
Catalog number FSA451050010
Price (MYR)
542.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
542.00
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage.
CAS7732-18-5