Quartz Wool
Thermo Scientific Chemicals
Quartz Wool
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number FSA451040100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS62637-93-8
IUPAC NameN,N-dimethylmethanamine oxide dihydrate
Molecular FormulaC3H13NO3
InChI KeyPGFPZGKEDZGJQZ-UHFFFAOYSA-N
SMILESO.O.C[N+](C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point94°C to 98°C
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks