Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Thermo Scientific Chemicals
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
FSA4507200252.5 L
Catalog number FSA450720025
Price (MYR)
1,179.00
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
1,179.00
Add to cart
Chemical Identifiers
CAS452-86-8
IUPAC Name4-methylbenzene-1,2-diol
Molecular FormulaC7H8O2
InChI KeyZBCATMYQYDCTIZ-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Melting point63°C to 68°C
Appearance (Color)White to brown
Infrared spectrumConforms
GC>=97.5 %