Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
Thermo Scientific Chemicals
Hydrogen chloride, technical, 4 to 6N solution in 2-propanol
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
FSA4507200101 L
Catalog number FSA450720010
Price (MYR)
507.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
507.00
Add to cart
Chemical Identifiers
CAS2338-18-3
IUPAC Name2,3-dihydro-1H-inden-2-amine hydrochloridyl
Molecular FormulaC9H11ClN
InChI KeyDLYRFJDJKGGVKT-UHFFFAOYSA-N
SMILES[Cl].NC1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder