Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number FSA450680050
Price (MYR)
758.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
758.00
Add to cart
Chemical Identifiers
CAS6937-96-8
IUPAC Name2,6-dimethoxy-4-methylbenzaldehyde
Molecular FormulaC10H12O3
InChI KeyVVBGEVFGPIUGNB-UHFFFAOYSA-N
SMILESCOC1=CC(C)=CC(OC)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Free acid (titration)≥1.5%
Melting Point (clear melt)88.0-97.0?C