Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number FSA450680010
Price (MYR)
271.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
271.00
Add to cart
Specifications
Chemical Name or MaterialCalcium carbonate
Melting Point825.0°C
CAS471-34-1
Quantity1 kg
EINECS Number207-439-9