Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass Bottle
Catalog number FSA450255000
Price (MYR)
2,058.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,058.00
Add to cart
Chemical Identifiers
CAS16919-58-7
IUPAC Nameplatinum(4+) diammonium hexachloride
Molecular FormulaCl6H8N2Pt
InChI KeyPCCGQTHFYHJATL-UHFFFAOYSA-J
SMILES[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 43.4% min