Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Metaphosphoric acid, Thermo Scientific Chemicals
CAS: 37267-86-0 | HO3P | 79.979 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Catalog number FSA450251000
Price (MYR)
629.00
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
629.00
Add to cart
Chemical Identifiers
CAS672-15-1
IUPAC Name(2S)-2-amino-4-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyUKAUYVFTDYCKQA-VKHMYHEASA-N
SMILESN[C@@H](CCO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Optical Rotation-8.6 ± 1° (c=2 in water)
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%