2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number FSA450182500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Colorless to light yellow to light blue-grey
Appearance (Form)Solution
Titration Argentometric0.9 to 1.1 M
Water=<0.2 % (Coulometric)