2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Iminothiolane hydrochloride, 98%, Thermo Scientific Chemicals
CAS: 4781-83-3 | C4H7NS·ClH | 137.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number FSA450180010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS513-49-5
IUPAC Namebutan-2-amine
Molecular FormulaC4H11N
InChI KeyBHRZNVHARXXAHW-UHFFFAOYNA-N
SMILESCCC(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Optical Rotation8.2 ± 0.3? (neat)
Assay (GC)≥97.5%
Refractive Index1.3910-1.3950 @ 20?C