Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA449550050
Price (MYR)
690.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
690.00
Add to cart
Chemical Identifiers
CAS4422-95-1
IUPAC Namebenzene-1,3,5-tricarbonyl trichloride
Molecular FormulaC9H3Cl3O3
InChI KeyUWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILESClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 % (ex Cl)
Infrared spectrumConforms
Melting point30°C to 35°C
Appearance (Form)Solid or liquid
Appearance (Color)White to yellow