Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Atenolol, 98%, Thermo Scientific Chemicals
CAS: 29122-68-7 | C14H22N2O3 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA449550010
Price (MYR)
388.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
388.00
Add to cart
Chemical Identifiers
CAS35225-79-7
IUPAC Name(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one
Molecular FormulaC17H14O
InChI KeyWMKGGPCROCCUDY-PHEQNACWSA-N
SMILESO=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point108°C to 114°C
Assay>=97.5 %