Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Cytidine 5'-monophosphate, 97%, from yeast
Thermo Scientific Chemicals
Cytidine 5'-monophosphate, 97%, from yeast
CAS: 63-37-6 | C9H14N3O8P | 323.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA449490050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.005mm (0.0002in) thick.
Assay from Supplier's CofA≥99.97% (metals basis)
Appearance (Color)Silver