1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, pure, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
25 L
Catalog number FSA449180050
Price (MYR)
805.00
Add to cart
Quantity:
5 L
Request bulk or custom format
Price (MYR)
805.00
Add to cart
Chemical Identifiers
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Assay (Titration)24.4 to 26.0 wt%
Chloride content10 ppm, max
Water Content (Karl Fischer Titration)0.1% max
Formliquid
Suitable for battery materials development.
RUO – Research Use Only