Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Alfa Aesar
Poly(acrylamide), carboxyl modified, high carboxyl content, approx. MW 100,000, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number FSA448532500
Price (MYR)
523.55
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
523.55
Add to cart
Chemical Identifiers
CAS626-15-3
IUPAC Name1,3-bis(bromomethyl)benzene
Molecular FormulaC8H8Br2
InChI KeyOXHOPZLBSSTTBU-UHFFFAOYSA-N
SMILESBrCC1=CC(CBr)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)71.0-81.0?C