Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid, 99+%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA448111000
Price (MYR)
365.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
365.00
Add to cart
Chemical Identifiers
CAS609-65-4
IUPAC Name2-chlorobenzoyl chloride
Molecular FormulaC7H4Cl2O
InChI KeyONIKNECPXCLUHT-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Titration RCOCl>=97.5 %
Infrared spectrumConforms
Refractive index1.5710 to 1.5750 (20°C, 589 nm)
Appearance (Form)Liquid