Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Thermo Scientific Chemicals
Sodium benzoate, 99%, for biochemistry
CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Catalog number FSA447802500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1665-00-5
IUPAC Namedichloro(²H₂)methane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-DICFDUPASA-N
SMILES[2H]C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.04 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.4 %