Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Thermo Scientific Chemicals
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA447362500250 gPlastic bottle
Catalog number FSA447362500
Price (MYR)
1,578.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,578.00
Add to cart
Chemical Identifiers
CAS12036-10-1
IUPAC Namedioxoruthenium
Molecular FormulaO2Ru
InChI KeyWOCIAKWEIIZHES-UHFFFAOYSA-N
SMILESO=[Ru]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Black
Assay from Supplier's CofA≥99.9% RuO₂
Elemental Analysis(Ruthenium, Ru) : 73.0-76.5%
For HPLC
RUO – Research Use Only